Assay Details
| Gene Symbol: Cxcl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013655.4 | NP_038683.1 | 3 - 4 | 383 | 83 | |
| GenBank mRNA | AK157553.1 | - | 3 - 4 | 344 | 83 | |
| BB610070.1 | - | 3 - 4 | 339 | 83 | ||
| D43805.1 | - | 3 - 4 | 340 | 83 | ||
| L12030.1 | - | 3 - 4 | 338 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cxcl12
Genomic Map