Assay Details
Gene Symbol: Slc22a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013667.2 | NP_038695.1 | 6 - 7 | 1156 | 77 | |
GenBank mRNA | AJ006036.1 | - | 6 - 7 | 1156 | 77 | |
AK052658.1 | - | 6 - 7 | 1153 | 77 | ||
AK143892.1 | - | 6 - 7 | 1157 | 77 | ||
BC015250.1 | - | 6 - 7 | 1084 | 4242038 | 77 | |
BC069911.1 | - | 6 - 7 | 1103 | 4974824 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
