Assay Details
| Gene Symbol: Plk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013807.2 | NP_038835.2 | 8 - 9 | 1170 | 89 | |
| GenBank mRNA | AF136586.1 | - | 1 - 2 | 87 | 89 | |
| AK156754.1 | - | 8 - 9 | 1182 | 89 | ||
| BC031180.1 | - | 8 - 9 | 1162 | 4988360 | 89 | |
| BC063051.1 | - | 8 - 9 | 1170 | 5718130 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plk3
Genomic Map