Assay Details
Gene Symbol: Abca8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013851.2 | NP_038879.2 | 3 - 4 | 505 | 69 | |
GenBank mRNA | AF498362.1 | - | 3 - 4 | 505 | 69 | |
AK138196.1 | - | 3 - 4 | 506 | 69 | ||
AK173034.1 | - | 3 - 4 | 432 | 69 | ||
BC141401.1 | - | 3 - 4 | 468 | 9056014 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
