Assay Details
Gene Symbol: Abca8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013851.2 | NP_038879.2 | 29 - 30 | 4031 | 61 | |
GenBank mRNA | AF498362.1 | - | 29 - 30 | 4031 | 61 | |
AK173034.1 | - | 28 - 29 | 3772 | 61 | ||
BC141401.1 | - | 29 - 30 | 3994 | 9056014 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
