Assay Details
Gene Symbol: Abca8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013851.2 | NP_038879.2 | 32 - 33 | 4405 | 101 | |
GenBank mRNA | AF498362.1 | - | 32 - 33 | 4405 | 101 | |
AK173034.1 | - | 31 - 32 | 4146 | 101 | ||
BC141401.1 | - | 32 - 33 | 4368 | 9056014 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
