Assay Details
| Gene Symbol: Prss30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013921.3 | NP_038949.2 | 2 - 3 | 122 | 86 | |
| GenBank mRNA | AJ243866.1 | - | 2 - 3 | 85 | 86 | |
| BC010970.1 | - | 2 - 3 | 115 | 4216644 | 86 | |
| BC040348.1 | - | 2 - 3 | 122 | 4206523 | 86 | |
More Information
Set Membership: |
|

Prss30
Genomic Map