Assay Details
| Gene Symbol: Oaz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016901.3 | NP_058597.2 | 1 - 2 | 264 | 86 | |
| GenBank mRNA | AB045835.1 | - | 1 - 2 | 189 | 86 | |
| AF175297.1 | - | 1 - 2 | 229 | 86 | ||
| AK006116.1 | - | 1 - 2 | 199 | 86 | ||
| AK006634.1 | - | 1 - 2 | 200 | 86 | ||
| AK076942.1 | - | 1 - 2 | 206 | 86 | ||
| BC119339.1 | - | 1 - 2 | 191 | 86 | ||
| BC119341.1 | - | 1 - 2 | 191 | 86 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF175297.1 transcripts.
Set Membership: |
|

Oaz3
Genomic Map