Assay Details
| Gene Symbol: Ywhab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018753.6 | NP_061223.2 | 3 - 4 | 598 | 119 | |
| GenBank mRNA | AF058797.1 | - | 3 - 4 | 597 | 119 | |
| AK002632.1 | - | 3 - 4 | 617 | 119 | ||
| AK004872.1 | - | 3 - 4 | 599 | 119 | ||
| AK011389.1 | - | 3 - 4 | 598 | 119 | ||
| AK083367.1 | - | 3 - 4 | 588 | 119 | ||
| AK144061.1 | - | 3 - 4 | 661 | 119 | ||
| AK150414.1 | - | 3 - 4 | 611 | 119 | ||
| AK151294.1 | - | 3 - 4 | 611 | 119 | ||
| AK158932.1 | - | 3 - 4 | 501 | 119 | ||
| AK178774.1 | - | 2 - 3 | 158 | 119 | ||
| AK219160.1 | - | 1 - 2 | 130 | 119 | ||
| BC049070.1 | - | 432 | 119 | |||
| BC132121.1 | - | 3 - 4 | 452 | 40130398 | 119 | |
| BC138512.1 | - | 3 - 4 | 452 | 8861532 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ywhab
Genomic Map