Assay Details
| Gene Symbol: Prss16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019429.2 | NP_062302.1 | 7 - 8 | 763 | 68 | |
| GenBank mRNA | AJ131775.1 | - | 7 - 8 | 763 | 68 | |
| AK088019.1 | - | 7 - 8 | 791 | 68 | ||
| BC125481.1 | - | 7 - 8 | 761 | 40129996 | 68 | |
| BC131955.1 | - | 7 - 8 | 761 | 40130232 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prss16
Genomic Map