Assay Details
| Gene Symbol: Unc93b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161428.1 | NP_001154900.1 | 5 - 6 | 757 | 114 | |
| NM_019449.2 | NP_062322.2 | 5 - 6 | 757 | 114 | ||
| GenBank mRNA | AK030488.1 | - | 5 - 6 | 718 | 114 | |
| AK155556.1 | - | 5 - 6 | 715 | 114 | ||
| AK162535.1 | - | 5 - 6 | 710 | 114 | ||
| AK165060.1 | - | 5 - 6 | 717 | 114 | ||
| BC018388.1 | - | 5 - 6 | 724 | 4209296 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc93b1
Genomic Map