Assay Details
| Gene Symbol: Scamp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019575.4 | NP_062521.1 | 2 - 3 | 111 | 81 | |
| GenBank mRNA | AF224721.1 | - | 2 - 3 | 86 | 81 | |
| AF295102.1 | - | 2 - 3 | 104 | 81 | ||
| AK019112.1 | - | 2 - 3 | 114 | 81 | ||
| AK078226.1 | - | 2 - 3 | 102 | 81 | ||
| AK154094.1 | - | 2 - 3 | 102 | 81 | ||
| AK160755.1 | - | 2 - 3 | 102 | 81 | ||
| AK168554.1 | - | 2 - 3 | 97 | 81 | ||
| BC018215.1 | - | 2 - 3 | 80 | 3965223 | 81 | |
| BC054104.2 | - | 2 - 3 | 106 | 30021104 | 81 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF224721.1,AF295102.1,AK154094.1,AK160755.1,AK168554.1,BC018215.1,BC054104.2,NM_019575.4 transcripts.
Set Membership: |
|

Scamp4
Genomic Map