Assay Details
| Gene Symbol: Wsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019653.3 | NP_062627.3 | 3 - 4 | 779 | 114 | |
| GenBank mRNA | AF033186.1 | - | 3 - 4 | 476 | 114 | |
| AK019206.2 | - | 3 - 4 | 756 | 114 | ||
| AK145974.1 | - | 3 - 4 | 758 | 114 | ||
| AK146752.1 | - | 3 - 4 | 754 | 114 | ||
| AK150394.1 | - | 3 - 4 | 754 | 114 | ||
| AK150600.1 | - | 3 - 4 | 754 | 114 | ||
| AK152545.1 | - | 3 - 4 | 757 | 114 | ||
| AK159905.1 | - | 3 - 4 | 754 | 114 | ||
| AK167174.1 | - | 3 - 4 | 756 | 114 | ||
| AK167394.1 | - | 3 - 4 | 757 | 114 | ||
| AK167942.1 | - | 3 - 4 | 754 | 114 | ||
| AK168468.1 | - | 3 - 4 | 757 | 114 | ||
| AK168565.1 | - | 3 - 4 | 757 | 114 | ||
| AK169058.1 | - | 3 - 4 | 754 | 114 | ||
| AK200135.1 | - | 1 - 2 | 238 | 114 | ||
| BC053391.1 | - | 3 - 4 | 530 | 30073116 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wsb1
Genomic Map