Assay Details
| Gene Symbol: Usp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038589.2 | NP_001033678.1 | 7 - 8 | 795 | 136 | |
| NM_021522.4 | NP_067497.2 | 8 - 9 | 900 | 136 | ||
| GenBank mRNA | AB034633.1 | - | 8 - 9 | 679 | 136 | |
| AK011322.1 | - | 8 - 9 | 836 | 136 | ||
| AK029977.1 | - | 8 - 9 | 829 | 136 | ||
| AK170933.1 | - | 894 | 136 | |||
| AK172477.1 | - | 8 - 9 | 887 | 136 | ||
| BC005571.1 | - | 8 - 9 | 819 | 3157723 | 136 | |
| BC050197.1 | - | 7 - 8 | 713 | 30006882 | 136 | |
| CK621440.1 | - | 2 - 3 | 121 | 136 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK170933.1 transcripts.
Set Membership: |
|

Usp14
Genomic Map