Assay Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022885.2 | NP_075023.2 | 8 - 9 | 1023 | 70 | |
| GenBank mRNA | AF233321.1 | - | 1 - 2 | 42 | 70 | |
| AF461761.1 | - | 8 - 9 | 922 | 70 | ||
| AK051183.1 | - | 8 - 9 | 1023 | 70 | ||
| AK090176.1 | - | 8 - 9 | 932 | 70 | ||
| AK166006.1 | - | 8 - 9 | 833 | 70 | ||
| BC029033.1 | - | 5 - 6 | 465 | 70 | ||
| BC029217.1 | - | 1 - 2 | 42 | 70 | ||
| BC033452.1 | - | 8 - 9 | 922 | 3156622 | 70 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF233321.1,BC029217.1 transcripts.
Set Membership: |
|

Slc30a5
Genomic Map