Assay Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022885.2 | NP_075023.2 | 10 - 11 | 1516 | 71 | |
| GenBank mRNA | AF233321.1 | - | 3 - 4 | 535 | 71 | |
| AF461761.1 | - | 10 - 11 | 1415 | 71 | ||
| AK051183.1 | - | 10 - 11 | 1516 | 71 | ||
| AK088004.1 | - | 9 - 10 | 1260 | 71 | ||
| AK090176.1 | - | 10 - 11 | 1425 | 71 | ||
| AK166006.1 | - | 10 - 11 | 1326 | 71 | ||
| AK183613.1 | - | 1 - 2 | 152 | 71 | ||
| BC029033.1 | - | 7 - 8 | 958 | 71 | ||
| BC033452.1 | - | 10 - 11 | 1415 | 3156622 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a5
Genomic Map