Assay Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022885.2 | NP_075023.2 | 14 - 15 | 2237 | 109 | |
| GenBank mRNA | AF233321.1 | - | 7 - 8 | 1256 | 109 | |
| AF461761.1 | - | 14 - 15 | 2136 | 109 | ||
| AK051183.1 | - | 14 - 15 | 2237 | 109 | ||
| AK088004.1 | - | 13 - 14 | 1981 | 109 | ||
| AK090176.1 | - | 14 - 15 | 2146 | 109 | ||
| AK166006.1 | - | 14 - 15 | 2047 | 109 | ||
| AK215365.1 | - | 1 - 2 | 193 | 109 | ||
| BC029033.1 | - | 11 - 12 | 1679 | 109 | ||
| BC029217.1 | - | 7 - 8 | 1089 | 109 | ||
| BC033452.1 | - | 14 - 15 | 2136 | 3156622 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a5
Genomic Map