Assay Details
| Gene Symbol: Grina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023168.3 | NP_075657.1 | 4 - 5 | 736 | 65 | |
| GenBank mRNA | AF182040.1 | - | 4 - 5 | 719 | 65 | |
| AK185141.1 | - | 1 - 2 | 65 | |||
| AK188174.1 | - | 1 - 2 | 37 | 65 | ||
| AK189399.1 | - | 3 - 4 | 360 | 65 | ||
| AK218437.1 | - | 1 - 2 | 37 | 65 | ||
| BC010802.1 | - | 4 - 5 | 769 | 4236429 | 65 | |
| BC019157.1 | - | 4 - 5 | 757 | 65 | ||
| BC037667.1 | - | 4 - 5 | 736 | 5376054 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grina
Genomic Map