Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300008929.mm.1 |
NM_023186.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008929.mm.1 |
NM_023186.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008929.mm.1 |
TC0300001102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008931.mm.1 |
NM_023186.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008931.mm.1 |
NM_023186.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008931.mm.1 |
TC0300001102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008934.mm.1 |
NM_023186.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008934.mm.1 |
NM_023186.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008934.mm.1 |
TC0300001102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008932.mm.1 |
NM_023186.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008932.mm.1 |
NM_023186.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008932.mm.1 |
TC0300001102.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008933.mm.1 |
NM_023186.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008933.mm.1 |
NM_023186.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008933.mm.1 |
TC0300001102.mm.1 |
MTA TC |