Assay Details
| Gene Symbol: Nop56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024193.2 | NP_077155.2 | 8 - 9 | 1071 | 71 | |
| GenBank mRNA | AK009799.1 | - | 8 - 9 | 1057 | 71 | |
| AK011481.1 | - | 8 - 9 | 1038 | 71 | ||
| AK077795.1 | - | 8 - 9 | 1038 | 71 | ||
| AK139924.1 | - | 8 - 9 | 1038 | 71 | ||
| AK146243.1 | - | 8 - 9 | 1038 | 71 | ||
| AK150258.1 | - | 8 - 9 | 1038 | 71 | ||
| BC002231.1 | - | 3 - 4 | 423 | 71 | ||
| BC021355.1 | - | 8 - 9 | 1033 | 5006404 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21628850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21628851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21628854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21628855 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21628852 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972263 | BC002231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187901 | BC002231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187901 | BC021355.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187901 | AK139924.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187901 | AK146243.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P187901 | AK150258.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105512 | AK030730.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105512 | AK037405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105512 | AK077795.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105512 | AK011744.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105512 | AK013528.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149319 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736069 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 280501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350987 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100215 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187901 | BC002231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187901 | BC021355.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187901 | AK139924.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187901 | AK146243.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P187901 | AK150258.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105512 | AK030730.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105512 | AK037405.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105512 | AK077795.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105512 | AK011744.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105512 | AK013528.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972263 | BC002231.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21628869 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100215 | - | - |
| Whole Mouse Genome G4122A | A_51_P187902 | BC021355.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187902 | AK030730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187902 | NM_024193.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P187902 | AI115137.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P370685 | AK154024.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P370685 | AK030730.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105341 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131366 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100215 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02016201 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02016202 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02016200 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02016195 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02016193 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01884851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21628855 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21628851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21628854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21628850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21628852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21628908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02096723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01981108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02016200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01981082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01981100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01981071 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02016195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02126557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01997679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02016201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01997678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02016202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01981094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21628834 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10476101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10476094 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426533_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455035_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10476093 | - | - |
| Mouse Genome 430 2.0 Array | 1455035_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1426533_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426533_at | - | - |
| Mouse Genome 430 2.0 Array | 1455035_PM_s_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200015629.mm.1 | NM_024193.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015629.mm.1 | NM_024193.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015629.mm.1 | TC0200001972.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200015634.mm.1 | NM_024193.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015634.mm.1 | NM_024193.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015634.mm.1 | TC0200001972.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200015648.mm.1 | NM_024193.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015648.mm.1 | NM_024193.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015648.mm.1 | TC0200001972.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200015650.mm.1 | NM_024193.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015650.mm.1 | NM_024193.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015650.mm.1 | TC0200001972.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200015628.mm.1 | NM_024193.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015628.mm.1 | NM_024193.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015628.mm.1 | TC0200001972.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200015643.mm.1 | NM_024193.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015643.mm.1 | NM_024193.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015643.mm.1 | TC0200001972.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200015627.mm.1 | NM_024193.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015627.mm.1 | NM_024193.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015627.mm.1 | TC0200001972.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200015616.mm.1 | NM_024193.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015616.mm.1 | NM_024193.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015616.mm.1 | TC0200001972.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200015651.mm.1 | NM_024193.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015651.mm.1 | NM_024193.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015651.mm.1 | TC0200001972.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200015642.mm.1 | NM_024193.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015642.mm.1 | NM_024193.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200015642.mm.1 | TC0200001972.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426533_at | - | - |
| Mouse Expression Set 430 | 1455035_PM_s_at | - | - |
| Mouse Expression Set 430 | 1426533_PM_at | - | - |
| Mouse Expression Set 430 | 1455035_s_at | - | - |
| Murine Genome U74v2 Set | 95109_PM_at | - | - |
| Murine Genome U74v2 Set | 95109_at | - | - |
| Murine Genome U74v2 Set | 162433_f_at | - | - |
| Murine Genome U74v2 Set | 162433_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455035_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455035_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426533_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426533_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426533_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426533_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455035_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455035_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5570139 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005570139 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245860 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245860 | - | - |
| mouse-6 v1.1 (Illumina) | scl067134.7_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5570139 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067134.7_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570139 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1245860 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245860 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067134.7_2-S | - | - |

Nop56
Genomic Map