Assay Details
Gene Symbol: Akr7a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025337.3 | NP_079613.3 | 4 - 5 | 725 | 83 | |
GenBank mRNA | AF525358.2 | - | 4 - 5 | 627 | 83 | |
AJ271800.1 | - | 4 - 5 | 642 | 83 | ||
AJ271801.1 | - | 4 - 5 | 642 | 83 | ||
AK003915.1 | - | 4 - 5 | 712 | 83 | ||
AK143203.1 | - | 4 - 5 | 724 | 83 | ||
BC031857.1 | - | 4 - 5 | 702 | 5124841 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
