Assay Details
| Gene Symbol: Asrgl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025610.3 | NP_079886.2 | 5 - 6 | 672 | 66 | |
| GenBank mRNA | AK006147.1 | - | 5 - 6 | 708 | 66 | |
| AK010381.1 | - | 5 - 6 | 714 | 66 | ||
| AK030209.1 | - | 5 - 6 | 673 | 66 | ||
| BC016106.1 | - | 3 - 4 | 424 | 4921265 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asrgl1
Genomic Map