Assay Details
| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025772.4 | NP_080048.2 | 6 - 7 | 608 | 74 | |
| GenBank mRNA | AJ404859.1 | - | 6 - 7 | 607 | 74 | |
| AK010924.1 | - | 2 - 3 | 220 | 74 | ||
| AK017400.1 | - | 2 - 3 | 386 | 74 | ||
| AK159656.1 | - | 6 - 7 | 733 | 74 | ||
| AY265461.1 | - | 2 - 3 | 386 | 74 | ||
| BC018350.1 | - | 6 - 7 | 599 | 4195234 | 74 | |
| BC048682.1 | - | 6 - 7 | 653 | 74 | ||
| BC058574.1 | - | 6 - 7 | 581 | 6487720 | 74 | |
More Information
Set Membership: |
|

Dtnbp1
Genomic Map