Assay Details
Gene Symbol: Prl3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025896.2 | NP_080172.1 | 1 - 2 | 185 | 70 | |
GenBank mRNA | AF525154.1 | - | 1 - 2 | 170 | 70 | |
AK005474.1 | - | 1 - 2 | 186 | 70 | ||
AK005567.1 | - | 1 - 2 | 183 | 70 | ||
BC100334.1 | - | 1 - 2 | 122 | 30791042 | 70 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF525154.1,AK005474.1,BC100334.1,NM_025896.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987326 | AK005474.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987326 | BC100334.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987326 | NM_025896.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621120 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32375 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 163830 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223450 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935413 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636576 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 517796 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54542 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127249 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 142888 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150358 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P127529 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987326 | AK005474.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987326 | BC100334.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987326 | NM_025896.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901504 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901497 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901499 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901492 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901469 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901498 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901502 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901470 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901484 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901486 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901500 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901495 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901488 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901477 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901474 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901481 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901479 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901476 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901503 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901506 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901491 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901496 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901471 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901501 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901475 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901490 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901478 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28901494 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P127529 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07818315 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150358 | - | - |
Whole Mouse Genome G4122A | A_51_P431347 | AF525154.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431347 | AK005474.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431347 | AK005567.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431347 | BC100334.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P431347 | NM_025896.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113259 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150358 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P127529 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11042704 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10621567 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10511765 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10511766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901498 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901500 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901467 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901569 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901566 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901541 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28901491 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10529738 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901494 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901484 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901451 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10813477 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11042711 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901480 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901506 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901488 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10885438 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901502 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901453 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901458 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901474 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901503 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10813476 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10813479 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901486 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901475 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901464 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901478 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901481 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10885441 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901498 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10885678 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901457 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901466 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901456 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901500 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901470 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901492 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10885676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10885432 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901461 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901479 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901499 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901490 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901495 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901491 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901471 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901462 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28901504 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10404304 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404306 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404307 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404303 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404305 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404308 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404302 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420746_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10404301 | - | - |
Mouse Genome 430 2.0 Array | 1420746_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1420746_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67000_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67000_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300001853.mm.1 | NM_025896.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001853.mm.1 | NM_025896.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001853.mm.1 | TC1300000298.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001855.mm.1 | NM_025896.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001855.mm.1 | NM_025896.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001855.mm.1 | TC1300000298.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001852.mm.1 | NM_025896.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001852.mm.1 | NM_025896.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001852.mm.1 | TC1300000298.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001854.mm.1 | NM_025896.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001854.mm.1 | NM_025896.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001854.mm.1 | TC1300000298.mm.1 | MTA TC |
Mouse Expression Set 430 | 1420746_at | - | - |
Mouse Expression Set 430 | 1420746_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1420746_at | - | - |
Mouse Genome 430A 2.0 Array | 1420746_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420746_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420746_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 438212 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6760538 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006760538 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2589586 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2589586 | - | - |
mouse-6 v1.1 (Illumina) | scl44965.5.1_0-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6760538 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl44965.5.1_0-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6760538 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2589586 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2589586 | - | - |
