Assay Details
Gene Symbol: Snrpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026095.4 | NP_080371.1 | 1 - 2 | 85 | 75 | |
GenBank mRNA | AK005341.1 | - | 1 - 2 | 107 | 75 | |
AK006692.1 | - | 1 - 2 | 85 | 75 | ||
AK160321.1 | - | 1 - 2 | 579 | 75 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK005341.1,AK006692.1,AK160321.1,NM_026095.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27256923 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27256922 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27256924 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008747 | AK160321.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008747 | NM_026095.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008747 | AK005341.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008747 | AK006692.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 900729 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416760 | AJ487854.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416760 | AK220417.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416760 | BC145695.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416760 | NM_153135.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838078 | AF330197.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838078 | AK036351.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838078 | AK048110.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838078 | AK137148.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838078 | AK142598.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838078 | AK164502.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838078 | AK169999.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838078 | BC011463.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838078 | BC026485.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838078 | BC054842.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838078 | BC069835.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838078 | NM_033604.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595844 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535005 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546478 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683217 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86573 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 640718 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532301 | BC004761.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89144 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 98905 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179423 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115468 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008747 | AK160321.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008747 | NM_026095.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008747 | AK005341.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008747 | AK006692.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256935 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256885 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256905 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256887 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256939 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256883 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256913 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256906 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256912 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256923 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256917 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256889 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256931 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256904 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256918 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256907 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256884 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256891 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256928 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256886 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256916 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256902 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256897 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256934 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256929 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256893 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256899 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256922 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256937 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256895 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256892 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256903 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256920 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256909 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256933 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256900 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256910 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256924 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256925 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27256921 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115468 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02161488 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07110533 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179423 | - | - |
Whole Mouse Genome G4122A | A_51_P176681 | BC011510.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P176681 | AK160321.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P176681 | NM_026095.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P176681 | AK005341.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P176681 | AK006692.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P523179 | BC011510.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P523179 | AK160321.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P523179 | NM_026095.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P523179 | AK005341.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P523179 | AK006692.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P150588 | AK163643.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122725 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179423 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115468 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08669000 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08668997 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27256922 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27256924 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27256923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27256961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27257021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256939 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256969 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08629446 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256925 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256935 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27257019 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256973 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08374706 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256945 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256958 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08230197 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08668994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256968 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27257028 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08602354 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256950 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27257029 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08839309 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08327098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08520133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08697310 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08333102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256974 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256961 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08251896 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256963 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08764309 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256944 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08669000 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256955 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256957 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08839307 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256928 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27257025 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256954 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27256938 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08587525 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10364055 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364054 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364053 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364052 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1422884_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1422885_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10364051 | - | - |
Mouse Genome 430 2.0 Array | 1449629_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1422884_at | - | - |
Mouse Genome 430 2.0 Array | 1449629_s_at | - | - |
Mouse Genome 430 2.0 Array | 1422885_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1422884_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1422885_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67332_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67332_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1000005084.mm.1 | NM_026095.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000005084.mm.1 | NM_026095.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000005084.mm.1 | TC1000000716.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000005083.mm.1 | NM_026095.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000005083.mm.1 | NM_026095.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000005083.mm.1 | TC1000000716.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000005085.mm.1 | NM_026095.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000005085.mm.1 | NM_026095.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000005085.mm.1 | TC1000000716.mm.1 | MTA TC |
Mouse Expression Set 430 | 1422885_at | - | - |
Mouse Expression Set 430 | 1422884_PM_at | - | - |
Mouse Expression Set 430 | 1422885_PM_at | - | - |
Mouse Expression Set 430 | 1422884_at | - | - |
Murine Genome U74v2 Set | 98077_PM_at | - | - |
Murine Genome U74v2 Set | 98077_at | - | - |
Murine Genome U74v2 Set | 167826_r_at | - | - |
Murine Genome U74v2 Set | 167826_PM_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1422885_at | - | - |
Mouse Genome 430A 2.0 Array | 1422884_at | - | - |
Mouse Genome 430A 2.0 Array | 1422884_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422885_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422884_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422884_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422885_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422885_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 669114 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2970050 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002970050 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244776 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1244776 | - | - |
mouse-6 v1.1 (Illumina) | scl067332.3_15-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2970050 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl067332.3_15-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2970050 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1244776 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244776 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067332.3_15-S | - | - |
