Assay Details
| Gene Symbol: Retsat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026159.4 | NP_080435.3 | 5 - 6 | 992 | 84 | |
| GenBank mRNA | AK002851.1 | - | 2 - 3 | 344 | 84 | |
| AK144115.1 | - | 1148 | 84 | |||
| AY704159.1 | - | 5 - 6 | 992 | 84 | ||
| BC011203.1 | - | 1 - 2 | 174 | 84 | ||
| BC117750.1 | - | 5 - 6 | 987 | 84 | ||
| BC117751.1 | - | 5 - 6 | 992 | 40090531 | 84 | |
| EF620363.1 | - | 1018 | 84 | |||
| EF620364.1 | - | 1018 | 84 | |||
| EF620365.1 | - | 5 - 6 | 1018 | 84 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK002851.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P319070 | AK002851.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P319070 | AK002851.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 845994 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72366 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447306 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410403 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153440 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P319070 | AK002851.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P319070 | AK002851.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P319070 | AK002851.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P319070 | AK002851.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24542539 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05939192 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153440 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05939178 | - | - |
| Whole Mouse Genome G4122A | A_51_P319070 | AK002851.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P319070 | AK002851.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105413 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153440 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05560888 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05560897 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05560895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24542644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05082036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05395571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05560888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05395570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05303878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05223225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05303874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05223230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05223228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05082038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05639523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24542541 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539150 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539153 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539144 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539147 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539146 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539148 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539149 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539151 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539145 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539152 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424716_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424715_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539143 | - | - |
| Mouse Genome 430 2.0 Array | 1424715_at | - | - |
| Mouse Genome 430 2.0 Array | 1424716_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424715_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424716_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67442_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67442_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600006568.mm.1 | TC0600000854.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600006568.mm.1 | NM_026159.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006568.mm.1 | NM_026159.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006569.mm.1 | TC0600000854.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600006569.mm.1 | NM_026159.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600006569.mm.1 | NM_026159.4 | Refseq |
| Mouse Expression Set 430 | 1424715_at | - | - |
| Mouse Expression Set 430 | 1424716_PM_at | - | - |
| Mouse Expression Set 430 | 1424715_PM_at | - | - |
| Mouse Expression Set 430 | 1424716_at | - | - |
| Murine Genome U74v2 Set | 95026_PM_at | - | - |
| Murine Genome U74v2 Set | 95026_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424715_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424715_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424716_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424716_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424716_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424715_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424716_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424715_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 872010 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 460110 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 460110 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4760603 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000460110 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004760603 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2769680 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240471 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240471 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2769680 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001019.1_73-S | - | - |
| mouse-6 v1.1 (Illumina) | scl29880.12.1_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4760603 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 460110 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001019.1_73-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29880.12.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 460110 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4760603 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2769680 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240471 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2769680 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240471 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl29880.12.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001019.1_73-S | - | - |

Retsat
Genomic Map