Assay Details
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026474.5 | NP_080750.1 | 1 - 2 | 175 | 107 | |
GenBank mRNA | AK010830.1 | - | 1 - 2 | 92 | 106 | |
AK020433.1 | - | 1 - 2 | 94 | 107 | ||
AK045153.1 | - | 1 - 2 | 114 | 107 | ||
AK082328.1 | - | 1 - 2 | 177 | 107 | ||
AK141416.1 | - | 111 | 107 | |||
AK148838.1 | - | 1 - 2 | 89 | 107 | ||
BC009167.1 | - | 1 - 2 | 92 | 2811924 | 107 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK020433.1,AK045153.1,AK082328.1,AK141416.1,AK148838.1,BC009167.1,NM_026474.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
