Assay Details
| Gene Symbol: Sugt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026474.5 | NP_080750.1 | 10 - 11 | 761 | 86 | |
| GenBank mRNA | AK007881.1 | - | 10 - 11 | 696 | 86 | |
| AK010711.1 | - | 9 - 10 | 730 | 86 | ||
| AK010830.1 | - | 10 - 11 | 678 | 86 | ||
| AK020433.1 | - | 10 - 11 | 680 | 86 | ||
| AK045153.1 | - | 10 - 11 | 700 | 86 | ||
| AK082328.1 | - | 10 - 11 | 763 | 86 | ||
| AK141416.1 | - | 697 | 86 | |||
| AK148838.1 | - | 10 - 11 | 675 | 86 | ||
| BC009167.1 | - | 10 - 11 | 678 | 2811924 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sugt1
Genomic Map