Assay Details
Gene Symbol: Etaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026576.3 | NP_080852.2 | 3 - 4 | 518 | 97 | |
GenBank mRNA | AK019965.1 | - | 3 - 4 | 519 | 97 | |
AK034138.1 | - | 3 - 4 | 511 | 97 | ||
BC028916.1 | - | 3 - 4 | 513 | 3676092 | 97 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK034138.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27618336 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27618338 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27618341 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475040 | BC028916.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475040 | AK019965.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475040 | AK034138.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475040 | NM_026576.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475040 | AK018594.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475040 | BC028916.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475040 | AK019965.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475040 | AK034138.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475040 | NM_026576.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475040 | AK018594.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511242 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692201 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364745 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381442 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92597 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928942 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913422 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64196 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164606 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 179322 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143582 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115808 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475040 | BC028916.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475040 | AK019965.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475040 | AK034138.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475040 | NM_026576.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475040 | AK018594.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475040 | BC028916.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475040 | AK019965.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475040 | AK034138.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475040 | NM_026576.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475040 | AK018594.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618326 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618335 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618324 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618363 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618350 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618331 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618329 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618337 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618348 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618360 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618336 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618340 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618345 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618356 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618321 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618333 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618358 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618353 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618351 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618327 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618343 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618339 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27618330 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02270285 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115808 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143582 | - | - |
Whole Mouse Genome G4122A | A_51_P518462 | AK034138.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P518462 | NM_026576.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475040 | BC028916.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475040 | AK019965.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475040 | AK034138.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475040 | NM_026576.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475040 | AK018594.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475040 | BC028916.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475040 | AK019965.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475040 | AK034138.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475040 | NM_026576.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475040 | AK018594.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P299228 | AK160833.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108489 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119555 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115808 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143582 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09411519 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09411534 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09715587 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27618341 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27618336 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27618338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618306 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27618293 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09511784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618327 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618304 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618336 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618337 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618309 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09411506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618320 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09537901 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618331 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618360 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09202247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618301 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618321 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618311 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09368118 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618310 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618365 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618369 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618352 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618316 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618294 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618340 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618350 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618348 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618367 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09411545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618362 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618346 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618303 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09200737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09368124 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09381141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618364 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618323 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618366 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618343 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09200765 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618353 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618293 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27618306 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10384488 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384487 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384491 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384492 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384489 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10384490 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1457882_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1453064_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10384486 | - | - |
Mouse Genome 430 2.0 Array | 1453064_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1457882_at | - | - |
Mouse Genome 430 2.0 Array | 1453064_at | - | - |
Mouse Genome 430 2.0 Array | 1457882_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68145_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68145_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100021803.mm.1 | NM_026576.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100021803.mm.1 | NM_026576.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100021803.mm.1 | TC1100002255.mm.1 | MTA TC |
Mouse Expression Set 430 | 1453064_PM_at | - | - |
Mouse Expression Set 430 | 1453064_at | - | - |
Mouse Expression Set 430 | 1457882_PM_at | - | - |
Mouse Expression Set 430 | 1457882_at | - | - |
Mouse Genome 430A 2.0 Array | 1453064_at | - | - |
Mouse Genome 430A 2.0 Array | 1453064_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1453064_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1453064_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457882_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457882_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 681554 | - | - |
ABI Mouse Genome Survey Microarray | 844674 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3800400 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 100430181 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3800400 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003800400 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240716 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1240716 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2520417 | - | - |
mouse-6 v1.1 (Illumina) | scl40497.6_233-S | - | - |
mouse-6 v1.1 (Illumina) | scl0068145.2_4-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 100430181 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3800400 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0068145.2_4-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100430181 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800400 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1240716 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2520417 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240716 | - | - |
