Assay Details
| Gene Symbol: Cmtm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027022.4 | NP_081298.1 | 4 - 5 | 631 | 75 | |
| GenBank mRNA | AK005603.1 | - | 4 - 5 | 584 | 75 | |
| AK161303.1 | - | 4 - 5 | 625 | 75 | ||
| AY241871.1 | - | 4 - 5 | 631 | 75 | ||
| BC049747.1 | - | 4 - 5 | 624 | 6772661 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056789 | BC049747.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056789 | AY241871.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056789 | NM_027022.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056789 | AK005603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056789 | AK161303.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056792 | AK006874.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317225 | AY241871.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317225 | NM_027022.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317225 | AK005603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317225 | AK161303.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317225 | BC049747.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317225 | AY241871.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317225 | NM_027022.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317225 | AK005603.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317225 | AK161303.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317225 | BC049747.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349406 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 688774 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562383 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432041 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816302 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 62939 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 148012 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056792 | AK006874.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317225 | AY241871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317225 | NM_027022.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317225 | AK005603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317225 | AK161303.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317225 | BC049747.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317225 | AY241871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317225 | NM_027022.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317225 | AK005603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317225 | AK161303.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317225 | BC049747.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056789 | BC049747.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056789 | AY241871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056789 | NM_027022.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056789 | AK005603.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056789 | AK161303.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317225 | AY241871.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317225 | NM_027022.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317225 | AK005603.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317225 | AK161303.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317225 | BC049747.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317225 | AY241871.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317225 | NM_027022.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317225 | AK005603.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317225 | AK161303.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317225 | BC049747.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26127786 | - | - |
| Whole Mouse Genome G4122A | A_51_P317225 | AY241871.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317225 | NM_027022.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317225 | AK005603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317225 | AK161303.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317225 | BC049747.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317225 | AY241871.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317225 | NM_027022.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317225 | AK005603.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317225 | AK161303.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317225 | BC049747.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125048 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07038072 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07038079 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07038082 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07038074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26127784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06883664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06737665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07025496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07025497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07186899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07038072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26127781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06691116 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581050 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581052 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581051 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581053 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451976_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442797_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426224_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581049 | - | - |
| Mouse Genome 430 2.0 Array | 1451976_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1426224_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1442797_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1451976_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1431710_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426224_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1431710_at | - | - |
| Mouse Genome 430 2.0 Array | 1442797_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73381_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73381_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800021379.mm.1 | NM_027022.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021379.mm.1 | NM_027022.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021379.mm.1 | TC0800002798.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800021380.mm.1 | NM_027022.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021380.mm.1 | NM_027022.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021380.mm.1 | TC0800002798.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1442797_x_at | - | - |
| Mouse Expression Set 430 | 1442797_PM_x_at | - | - |
| Murine Genome U74v2 Set | 167347_PM_i_at | - | - |
| Murine Genome U74v2 Set | 167347_i_at | - | - |
| Murine Genome U74v2 Set | 168593_PM_r_at | - | - |
| Murine Genome U74v2 Set | 168593_r_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451976_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451976_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426224_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426224_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442797_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442797_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4920348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920348 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214978 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214978 | - | - |
| mouse-6 v1.1 (Illumina) | scl34423.9.1_49-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34423.9.1_49-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920348 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1214978 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214978 | - | - |

Cmtm2a
Genomic Map