Assay Details
Gene Symbol: Rufy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027425.3 | NP_081701.2 | 15 - 16 | 1659 | 76 | |
GenBank mRNA | AK173195.1 | - | 15 - 16 | 1620 | 76 | |
BC137736.1 | - | 15 - 16 | 1608 | 8860755 | 76 | |
BC137737.1 | - | 15 - 16 | 1608 | 8860756 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
