Assay Details
| Gene Symbol: Ldhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027570.3 | NP_081846.3 | 2 - 3 | 200 | 59 | |
| GenBank mRNA | AK014649.1 | - | 2 - 3 | 215 | 59 | |
| AK029820.1 | - | 214 | 59 | |||
| AY092768.1 | - | 2 - 3 | 205 | 59 | ||
| BC039155.1 | - | 2 - 3 | 196 | 4242682 | 59 | |
| BC055443.1 | - | 2 - 3 | 200 | 4235345 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ldhd
Genomic Map