Assay Details
| Gene Symbol: Ldhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027570.3 | NP_081846.3 | 4 - 5 | 476 | 62 | |
| GenBank mRNA | AK014649.1 | - | 4 - 5 | 491 | 62 | |
| AY092768.1 | - | 4 - 5 | 481 | 62 | ||
| BC039155.1 | - | 4 - 5 | 472 | 4242682 | 62 | |
| BC055443.1 | - | 4 - 5 | 476 | 4235345 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ldhd
Genomic Map