Assay Details
Gene Symbol: Cic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF363690.1 | - | 4 - 5 | 621 | 80 | |
AK147276.1 | - | 4 - 5 | 1254 | 80 | ||
BC058665.1 | - | 1 - 2 | 139 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
