Assay Details
| Gene Symbol: Zfp131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011832.1 | - | 2 - 3 | 174 | 72 | |
| AK030398.1 | - | 2 - 3 | 373 | 72 | ||
| AK047429.1 | - | 1 - 2 | 156 | 72 | ||
| AK134257.1 | - | 2 - 3 | 173 | 72 | ||
| AK135723.1 | - | 1 - 2 | 157 | 72 | ||
| AK162903.1 | - | 1 - 2 | 596 | 72 | ||
| AY092766.1 | - | 1 - 2 | 265 | 72 | ||
| BC023762.1 | - | 2 - 3 | 443 | 72 | ||
| BC057991.1 | - | 2 - 3 | 214 | 5323559 | 72 | |
| BE994245.1 | - | 1 - 2 | 315 | 72 | ||
| EF410128.1 | - | 398 | 72 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp131
Genomic Map