Assay Details
| Gene Symbol: Bmper | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028472.2 | NP_082748.1 | 5 - 6 | 1057 | 68 | |
| GenBank mRNA | AF454954.1 | - | 5 - 6 | 1057 | 68 | |
| AK014221.1 | - | 3 - 4 | 263 | 68 | ||
| AY263358.1 | - | 5 - 6 | 578 | 68 | ||
| BC066153.1 | - | 5 - 6 | 562 | 30098453 | 68 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK014221.1 transcripts.
Set Membership: |
|

Bmper
Genomic Map