Assay Details
| Gene Symbol: Bmper | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028472.2 | NP_082748.1 | 8 - 9 | 1350 | 101 | |
| GenBank mRNA | AF454954.1 | - | 8 - 9 | 1350 | 101 | |
| AK014221.1 | - | 6 - 7 | 555 | 101 | ||
| AY263358.1 | - | 8 - 9 | 871 | 101 | ||
| BC066153.1 | - | 8 - 9 | 855 | 30098453 | 101 | |
More Information
Set Membership: |
|

Bmper
Genomic Map