Assay Details
Gene Symbol: Prl8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028477.2 | NP_082753.1 | 1 - 2 | 90 | 107 | |
GenBank mRNA | AK005485.1 | - | 1 - 2 | 91 | 107 | |
AK014414.1 | - | 1 - 2 | 91 | 107 | ||
BC109185.1 | - | 1 - 2 | 89 | 40050251 | 107 | |
BC109186.1 | - | 1 - 2 | 89 | 40050253 | 107 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK005485.1,AK014414.1,BC109185.1,BC109186.1,NM_028477.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P565496 | AK014414.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P565496 | BC109185.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P565496 | BC109186.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P565496 | NM_028477.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P565496 | AF525158.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P565496 | AK014414.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P565496 | BC109185.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P565496 | BC109186.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P565496 | NM_028477.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P565496 | AF525158.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560441 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240333 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919235 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109550 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477821 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164309 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157830 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P565496 | AK014414.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P565496 | BC109185.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P565496 | BC109186.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P565496 | NM_028477.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P565496 | AF525158.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P565496 | AK014414.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P565496 | BC109185.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P565496 | BC109186.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P565496 | NM_028477.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P565496 | AF525158.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902725 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902724 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902731 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902734 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902756 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902744 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902751 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902730 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902739 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902760 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902727 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902729 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902753 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902722 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902742 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902726 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902733 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902747 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902755 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07818857 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157830 | - | - |
Whole Mouse Genome G4122A | A_52_P64459 | AK014414.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P565496 | AK014414.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P565496 | BC109185.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P565496 | BC109186.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P565496 | NM_028477.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P565496 | AF525158.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P565496 | AK014414.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P565496 | BC109185.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P565496 | BC109186.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P565496 | NM_028477.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P565496 | AF525158.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123307 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157830 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11108716 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11108744 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11108722 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11108731 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11108738 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11108739 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11108746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902676 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902772 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902744 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902726 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902739 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902756 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10576446 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902722 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902718 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902698 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902761 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902712 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902760 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902699 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902759 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902704 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902727 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902734 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902752 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10598433 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902757 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902707 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902714 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902747 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10576453 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902705 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902729 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902700 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902746 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902724 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902741 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902751 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902702 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902736 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902720 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902695 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902711 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902694 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902706 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902755 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902753 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902742 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902745 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902715 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10408406 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408407 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408409 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408408 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408404 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408410 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408411 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408403 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1453969_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432455_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408402 | - | - |
Mouse Genome 430 2.0 Array | 1453969_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1453969_at | - | - |
Mouse Genome 430 2.0 Array | 1432455_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1432455_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73244_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73244_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300010535.mm.1 | NM_028477.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010535.mm.1 | NM_028477.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010535.mm.1 | TC1300001693.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300010534.mm.1 | NM_028477.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010534.mm.1 | NM_028477.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010534.mm.1 | TC1300001693.mm.1 | MTA TC |
Mouse Expression Set 430 | 1432455_a_at | - | - |
Mouse Expression Set 430 | 1453969_PM_at | - | - |
Mouse Expression Set 430 | 1453969_at | - | - |
Mouse Expression Set 430 | 1432455_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432455_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432455_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453969_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453969_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 445460 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1410452 | - | - |
MouseWG-6 v2 Expression BeadChip | 102190750 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102190750 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1410452 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001410452 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217895 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1217895 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1213214 | - | - |
mouse-6 v1.1 (Illumina) | scl44169.6.1_114-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102190750 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1410452 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl44169.6.1_114-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1410452 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102190750 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1217895 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217895 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1213214 | - | - |
