Assay Details
| Gene Symbol: Bicd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039179.2 | NP_001034268.1 | 4 - 5 | 1193 | 76 | |
| NM_001039180.2 | NP_001034269.1 | 5 - 6 | 1089 | 76 | ||
| NM_029791.4 | NP_084067.1 | 4 - 5 | 1193 | 76 | ||
| GenBank mRNA | AJ250106.1 | - | 4 - 5 | 1152 | 76 | |
| AK122348.1 | - | 4 - 5 | 1166 | 76 | ||
| AK156407.1 | - | 4 - 5 | 1148 | 76 | ||
| AK171300.1 | - | 5 - 6 | 1047 | 76 | ||
| BC032198.1 | - | 4 - 5 | 1150 | 5324113 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bicd2
Genomic Map