Assay Details
| Gene Symbol: Slco2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033314.3 | NP_201571.2 | 6 - 7 | 988 | 64 | |
| GenBank mRNA | AF323958.1 | - | 6 - 7 | 857 | 64 | |
| AK087350.1 | - | 6 - 7 | 994 | 64 | ||
| AK166724.1 | - | 6 - 7 | 989 | 64 | ||
| AK169706.1 | - | 6 - 7 | 991 | 64 | ||
| BC035200.1 | - | 978 | 3158233 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slco2a1
Genomic Map