Assay Details
| Gene Symbol: Trim16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053169.2 | NP_444399.2 | 3 - 4 | 1075 | 65 | |
| GenBank mRNA | AF449496.1 | - | 3 - 4 | 826 | 65 | |
| AK053139.1 | - | 3 - 4 | 889 | 65 | ||
| AK078902.1 | - | 1 - 2 | 411 | 65 | ||
| BC052821.1 | - | 4 - 5 | 777 | 30057830 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim16
Genomic Map