Assay Details
| Gene Symbol: Selm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053267.2 | NP_444497.1 | 1 - 2 | 371 | 80 | |
| GenBank mRNA | AK018791.1 | - | 1 - 2 | 207 | 80 | |
| AK035312.1 | - | 1 - 2 | 161 | 80 | ||
| AK046034.1 | - | 1 - 2 | 163 | 80 | ||
| AK079358.1 | - | 1 - 2 | 189 | 80 | ||
| AK079576.1 | - | 1 - 2 | 390 | 80 | ||
| AY043488.1 | - | 1 - 2 | 187 | 80 | ||
| BC147332.1 | - | 1 - 2 | 281 | 9056406 | 80 | |
| BC147333.1 | - | 1 - 2 | 283 | 9056396 | 80 | |
| BI731890.1 | - | 1 - 2 | 79 | |||
| CK787580.1 | - | 1 - 2 | 363 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27552432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27552431 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P357133 | AK046034.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P357133 | AK079358.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P357133 | AK079576.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P357133 | AW049879.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P357133 | AY043488.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P357133 | AK046034.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P357133 | AK079358.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P357133 | AK079576.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P357133 | AW049879.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P357133 | AY043488.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937420 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171269 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P357133 | AK046034.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P357133 | AK079358.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P357133 | AK079576.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P357133 | AW049879.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P357133 | AY043488.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P357133 | AK046034.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P357133 | AK079358.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P357133 | AK079576.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P357133 | AW049879.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P357133 | AY043488.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P241601 | AK086243.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27552423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27552414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27552429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27552430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27552418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27552424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27552420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27552433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27552427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27552415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27552412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27552419 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171269 | - | - |
| Whole Mouse Genome G4122A | A_51_P241601 | AK086243.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P357133 | AK046034.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P357133 | AK079358.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P357133 | AK079576.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P357133 | AW049879.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P357133 | AY043488.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P357133 | AK046034.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P357133 | AK079358.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P357133 | AK079576.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P357133 | AW049879.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P357133 | AY043488.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137008 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171269 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09468902 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09468895 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09468898 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27552432 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27552431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27552385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09468902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09407221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09314625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09277239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09468897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09314615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09637580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09305565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27552419 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373772 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373775 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373777 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373771 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373774 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373773 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373769 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373776 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373770 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424394_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373768 | - | - |
| Mouse Genome 430 2.0 Array | 1424394_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424394_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 114679_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 114679_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100000240.mm.1 | NM_053267.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000240.mm.1 | TC1100000015.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100000241.mm.1 | NM_053267.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000241.mm.1 | TC1100000015.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100000242.mm.1 | NM_053267.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000242.mm.1 | TC1100000015.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100000238.mm.1 | NM_053267.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000238.mm.1 | TC1100000015.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424394_at | - | - |
| Mouse Expression Set 430 | 1424394_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424394_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424394_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424394_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424394_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 910715 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 450458 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 450458 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103170484 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000450458 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2707155 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2707155 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236263 | - | - |
| mouse-6 v1.1 (Illumina) | scl41904.3_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103170484 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 450458 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41904.3_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103170484 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 450458 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1226838 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226838 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2582503 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl41904.3_0-S | - | - |

Selm
Genomic Map