Assay Details
| Gene Symbol: Hist2h3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC015270.1 | - | 1 | 4240748 | 114 | |
| CO040940.1 | - | 1 | 340 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2142251 | NM_054045.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2142251 | BC015270.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142251 | NM_054045.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142251 | BC015270.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348322 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04997256 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04997256 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02709729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02653277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03172954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348294 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02966250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02653272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348332 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10500328 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494400 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494406 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494403 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422155_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10494405 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10494402 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10500327 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10494395 | - | - |
| Mouse Genome 430 2.0 Array | 1422155_at | - | - |
| Mouse Genome 430 2.0 Array | 1422155_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 97114_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 97114_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300019519.mm.1 | NM_054045.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019519.mm.1 | TC0300002500.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418367_PM_x_at | - | - |
| Mouse Expression Set 430 | 1460314_s_at | - | - |
| Mouse Expression Set 430 | 1460314_PM_s_at | - | - |
| Mouse Expression Set 430 | 1418367_x_at | - | - |
| Mouse Expression Set 430 | 1418366_at | - | - |
| Mouse Expression Set 430 | 1427864_PM_at | - | - |
| Mouse Expression Set 430 | 1453573_at | - | - |
| Mouse Expression Set 430 | 1427864_at | - | - |
| Mouse Expression Set 430 | 1453573_PM_at | - | - |
| Mouse Expression Set 430 | 1418366_PM_at | - | - |
| Mouse Expression Set 430 | 1422155_PM_at | - | - |
| Mouse Expression Set 430 | 1422155_at | - | - |
| Mouse Expression Set 430 | 1442051_PM_at | - | - |
| Mouse Expression Set 430 | 1442051_at | - | - |
| Murine Genome U74v2 Set | 93067_PM_f_at | - | - |
| Murine Genome U74v2 Set | 93067_f_at | - | - |
| Murine Genome U74v2 Set | 93023_PM_f_at | - | - |
| Murine Genome U74v2 Set | 93068_PM_r_at | - | - |
| Murine Genome U74v2 Set | 94756_PM_at | - | - |
| Murine Genome U74v2 Set | 96416_f_at | - | - |
| Murine Genome U74v2 Set | 93022_PM_i_at | - | - |
| Murine Genome U74v2 Set | 93022_i_at | - | - |
| Murine Genome U74v2 Set | 93068_r_at | - | - |
| Murine Genome U74v2 Set | 94756_at | - | - |
| Murine Genome U74v2 Set | 96416_PM_f_at | - | - |
| Murine Genome U74v2 Set | 93023_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453573_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418366_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453573_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422155_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418367_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427864_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422155_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418367_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418366_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460314_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460314_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427864_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422155_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422155_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 548057 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3990670 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3990670 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003990670 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244200 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2724358 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244200 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2724358 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3990670 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21816.1.35_40-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3990670 | - | - |

Hist2h3c2
Genomic Map