Assay Details
Gene Symbol: Wdr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080848.2 | NP_543124.1 | 5 - 6 | 572 | 80 | |
GenBank mRNA | AF416510.1 | - | 5 - 6 | 577 | 80 | |
AK075937.1 | - | 5 - 6 | 515 | 80 | ||
AK144091.1 | - | 5 - 6 | 516 | 80 | ||
BC016103.1 | - | 5 - 6 | 507 | 4919781 | 80 | |
BC025801.1 | - | 5 - 6 | 549 | 5132525 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
