Assay Details
| Gene Symbol: Vps4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126165.1 | NP_569053.1 | 6 - 7 | 764 | 66 | |
| GenBank mRNA | AK047821.1 | - | 6 - 7 | 776 | 66 | |
| AK159117.1 | - | 6 - 7 | 790 | 66 | ||
| AK169949.1 | - | 6 - 7 | 752 | 66 | ||
| BC018368.1 | - | 6 - 7 | 764 | 3963907 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vps4a
Genomic Map