Assay Details
| Gene Symbol: Cd209a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133238.5 | NP_573501.1 | 5 - 6 | 529 | 83 | |
| GenBank mRNA | AF373408.1 | - | 5 - 6 | 476 | 83 | |
| AF374470.1 | - | 4 - 5 | 395 | 83 | ||
| AK054139.1 | - | 5 - 6 | 493 | 83 | ||
| AK171639.1 | - | 5 - 6 | 473 | 83 | ||
| AY049062.1 | - | 5 - 6 | 529 | 83 | ||
| BC111095.1 | - | 4 - 5 | 577 | 40040036 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
regulation of gene expression
regulation of blood coagulation
regulation of T cell proliferation
viral entry into host cell
virus receptor activity
mannose binding
carbohydrate binding
metal ion binding
calcium-dependent protein binding
carbohydrate derivative binding
endocytosis
regulation of gene expression
regulation of blood coagulation
regulation of T cell proliferation
viral entry into host cell
virus receptor activity
mannose binding
carbohydrate binding
metal ion binding
calcium-dependent protein binding
carbohydrate derivative binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133884 | AK171639.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133884 | AY049062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133884 | BC111095.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133884 | AF373408.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133884 | AK054139.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133884 | AK171639.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133884 | AY049062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133884 | BC111095.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133884 | AF373408.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P133884 | AK054139.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018061 | AK171639.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018061 | AY049062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018061 | AF373408.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018061 | AK054139.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018061 | NM_133238.5 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 638844 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 370535 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584667 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116773 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018061 | AK171639.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018061 | AY049062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018061 | AF373408.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018061 | AK054139.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018061 | NM_133238.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133884 | AK171639.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133884 | AY049062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133884 | BC111095.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133884 | AF373408.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133884 | AK054139.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133884 | AK171639.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133884 | AY049062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133884 | BC111095.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133884 | AF373408.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P133884 | AK054139.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P133884 | AK171639.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P133884 | AY049062.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P133884 | BC111095.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P133884 | AF373408.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P133884 | AK054139.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P133884 | AK171639.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P133884 | AY049062.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P133884 | BC111095.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P133884 | AF373408.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P133884 | AK054139.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25615457 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116773 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06408241 | - | - |
| Whole Mouse Genome G4122A | A_51_P133884 | AK171639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133884 | AY049062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133884 | BC111095.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133884 | AF373408.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133884 | AK054139.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133884 | AK171639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133884 | AY049062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133884 | BC111095.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133884 | AF373408.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133884 | AK054139.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107777 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136320 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116773 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07073464 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07157531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07157537 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07157535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25615466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07157534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07073464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07427677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07225554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07151882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06950199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06744413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07341403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25615378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07289894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07144566 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10576790 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576793 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576786 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576792 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576794 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576788 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576791 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576789 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576787 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10576785 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426172_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10576784 | - | - |
| Mouse Genome 430 2.0 Array | 1426172_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1426172_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170786_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170786_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800013319.mm.1 | NM_133238.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800013319.mm.1 | NM_133238.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800013319.mm.1 | TC0800001619.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426172_a_at | - | - |
| Mouse Expression Set 430 | 1426172_PM_a_at | - | - |
| Murine Genome U74v2 Set | 134733_PM_r_at | - | - |
| Murine Genome U74v2 Set | 134733_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426172_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426172_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426172_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426172_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 420071 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6590113 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6590113 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006590113 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2617210 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2617210 | - | - |
| mouse-6 v1.1 (Illumina) | scl35152.7.1_21-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6590113 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl35152.7.1_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6590113 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2617210 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2617210 | - | - |

Cd209a
Genomic Map