| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300012367.mm.1 |
NM_001076676.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300012367.mm.1 |
NM_001076676.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300012367.mm.1 |
NM_001252486.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300012367.mm.1 |
NM_001252486.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300012367.mm.1 |
NM_133247.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300012367.mm.1 |
NM_133247.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300012367.mm.1 |
TC0300001530.mm.1 |
MTA TC |