Assay Details
| Gene Symbol: Slc30a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252478.1 | NP_001239407.1 | 10 - 11 | 743 | 92 | |
| NM_144798.6 | NP_659047.2 | 10 - 11 | 728 | 92 | ||
| GenBank mRNA | AF233346.1 | - | 9 - 10 | 680 | 92 | |
| AF395840.1 | - | 9 - 10 | 670 | 92 | ||
| AK082907.1 | - | 10 - 11 | 705 | 92 | ||
| AK150342.1 | - | 10 - 11 | 704 | 92 | ||
| AK151120.1 | - | 10 - 11 | 704 | 92 | ||
| AK160903.1 | - | 10 - 11 | 732 | 92 | ||
| BC005753.1 | - | 2 - 3 | 191 | 92 | ||
| BC066162.1 | - | 10 - 11 | 695 | 30471027 | 92 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC005753.1 transcripts.
Set Membership: |
|

Slc30a6
Genomic Map