Assay Details
| Gene Symbol: Camk1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144817.2 | NP_659066.1 | 5 - 6 | 578 | 120 | |
| GenBank mRNA | AF428262.1 | - | 5 - 6 | 526 | 120 | |
| AY212936.1 | - | 4 - 5 | 458 | 120 | ||
| BC021840.1 | - | 5 - 6 | 527 | 4502479 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camk1g
Genomic Map