Assay Details
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271353.1 | NP_001258282.1 | 4 - 5 | 689 | 81 | |
NM_144855.3 | NP_659104.1 | 4 - 5 | 692 | 81 | ||
NM_178224.3 | NP_835742.1 | 4 - 5 | 692 | 81 | ||
GenBank mRNA | AK077669.1 | - | 4 - 5 | 653 | 81 | |
AK085692.1 | - | 4 - 5 | 654 | 81 | ||
BC013472.1 | - | 4 - 5 | 618 | 4221477 | 81 | |
BC013480.1 | - | 4 - 5 | 628 | 4239757 | 81 | |
BC022144.1 | - | 4 - 5 | 653 | 81 | ||
BC026595.1 | - | 4 - 5 | 625 | 4974777 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
